missing translation for 'onlineSavingsMsg'
Learn More

Bis(trimethylsilyl)acetylene, 99%, ACROS Organics™

Brand:  Acros Organics 182012500

Code : NEW

Additional Details : CAS Number : 14630-40-1 Weight : 0.25000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : II


Packaging Glass bottle
Quantity 250g

 View more versions of this product

Product Code. 15119223

  • £440.00 / 250g


Estimated Shipment
18-07-2018
Add to basket


Specifications

Chemical Identifiers

Bis(trimethylsilyl)acetylene
14630-40-1
99%
(CH3)3SiC=CSi(CH3)3
MFCD00008276
99%
99%
C8H18Si2
170.41
BTMSA

Specifications

Bis(trimethylsilyl)acetylene
14630-40-1
Authentic
Liquid After Melting
99%
C8H18Si2
170.41
BTMSA
21.0° to 24.0°C
250g
99%
0.7500g/mL
Glass bottle
99%
1.4260 to 1.4280
(CH3)3SiC=CSi(CH3)3
MFCD00008276
Clear colorless
136.0° to 137.0°C
2°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bis(trimethylsilyl)acetylene

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.