missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(triethyltin)sulfate, 98%

Brand:  Alfa Aesar™ 039622.06

Code : NEW

Additional Details : CAS Number : 57-52-3 Weight : 0.00500kg Transport : UN number : 3146 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11364567

  • £115.54 / 5g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(triethyltin)sulfate
507.81
UN3146
57-52-3
MFCD00048509

Specifications

Bis(triethyltin)sulfate
57-52-3
Crystalline
507.81
UN3146
>300°C
5g
98%
MFCD00048509
Videos
SDS