missing translation for 'onlineSavingsMsg'
Learn More

Bis(tri-tert-butylphosphine)palladium(0), Thermo Scientific Chemicals

Catalog Number p-7059555
Click to view available options
Quantity:
0.25 g
1 g
53199-31-8
C24H54P2Pd
511.06
MFCD03094580
MXQOYLRVSVOCQT-UHFFFAOYSA-N
bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
2734558
palladium;tritert-butylphosphane
[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C
53199-31-8
C24H54P2Pd
511.06
MFCD03094580
MXQOYLRVSVOCQT-UHFFFAOYSA-N
bis tri-tert-butylphosphine palladium 0, bis tri-t-butylphosphine palladium 0, bis tri-tert-butylphosphine palladium, bis tri-tert-butylphosphane palladium, palladium; tritert-butylphosphane, palladium, bis tris 1,1-dimethylethyl phosphine, pd p tbu 3 2, pd t-bu3p 2, bis tri-t-butylphosphine palladium, di tri-tert-butylphosphine palladium 0
2734558
palladium;tritert-butylphosphane
[Pd].CC(C)(C)P(C(C)(C)C)C(C)(C)C.CC(C)(C)P(C(C)(C)C)C(C)(C)C

Coupling reactions, Heck couplings

Bis(tri-tert-butylphosphine)palladium(0) is used in Coupling reactions, Heck couplings. It acts as a palladium catalyst for reductive carbonylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under inert atmosphere
TRUSTED_SUSTAINABILITY
Melting Point 258°C to 272°C
Color White
Quantity 0.25 g
Sensitivity Air and light sensitive
Solubility Information Soluble in toluene benzene,slight soluble in chloroform and acetone. Decomposes in water.
Formula Weight 511.06
Concentration or Composition (by Analyte or Components) Pd 20.9%
Physical Form Powder
Chemical Name or Material Bis (tri-tert-butylphosphine)palladium (0)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'