missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)zirconium dichloride, 99%

Brand:  Alfa Aesar™ 039555.02

Code : NEW

Additional Details : CAS Number : 54039-38-2 Weight : 0.00050kg Transport : UN number : 3096 Chem class : 1 Pack group : II

Quantity 0.5g

 View more versions of this product

Product Code. 11328368

  • £76.70 / 0.50g


Estimated Shipment date
from Supplier 25-04-2018
Add to basket

Specifications

Chemical Identifiers

Bis(pentamethylcyclopentadienyl)zirconium dichloride
432.56
UN3096
54039-38-2
MFCD00058849
Decamethylzirconocene dichloride

Specifications

Bis(pentamethylcyclopentadienyl)zirconium dichloride
Ampule
99%
MFCD00058849
Decamethylzirconocene dichloride
0.5g
54039-38-2
Crystalline
432.56
UN3096
Pale yellow
Air & Moisture Sensitive
Videos
SDS