missing translation for 'onlineSavingsMsg'
Learn More

Bis(pentamethylcyclopentadienyl)nickel, Thermo Scientific Chemicals

Catalog Number p-7050662
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
74507-63-4
C20H30Ni
329.153
MFCD00058704
CHPLEWYRKUFKQP-UHFFFAOYSA-N
Decamethylnickelocene
24943158
nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]
This item is not returnable. View return policy
74507-63-4
C20H30Ni
329.153
MFCD00058704
CHPLEWYRKUFKQP-UHFFFAOYSA-N
Decamethylnickelocene
24943158
nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Ampule
Quantity 250 mg
Sensitivity Air and moisture sensitive
Formula Weight 329.17
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)nickel

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'