missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(pentamethylcyclopentadienyl)chromium, 96%

Brand:  Alfa Aesar™ 033653.04

Code : NEW

Additional Details : CAS Number : 74507-61-2 Weight : 0.00200kg Transport : UN number : 2846 Chem class : 1 Pack group : I

Quantity 2g

 View more versions of this product

Product Code. 11357768

  • £220.16 / 2g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(pentamethylcyclopentadienyl)chromium
322.45
Decamethylchromocene
74507-61-2
MFCD00058886

Specifications

Bis(pentamethylcyclopentadienyl)chromium
Powder
322.45
Decamethylchromocene
Air & Moisture Sensitive
74507-61-2
96%
MFCD00058886
2g
Videos
SDS