missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(pentafluorophenyl) carbonate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.
Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.
Notes
Incompatible with oxidizing agents.
Bis(pentafluorophenyl) carbonate is a reagent used as a carbonyl equivalent and for various coupling reactions, especially for azapeptides.
Solubility
Soluble in chloroform, ethyl acetate and methylene chloride.
Notes
Incompatible with oxidizing agents.
Specifications
Melting Point | 47°C to 50°C |
Flash Point | 110°C (230°F) |
Linear Formula | (C6F5O)2CO |
Quantity | 1 g |
Beilstein | 2029727 |
Solubility Information | Soluble in chloroform,ethyl acetate and methylene chloride. |
Formula Weight | 394.12 |
Percent Purity | ≥98% |
Chemical Name or Material | Bis(pentafluorophenyl) carbonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'