missing translation for 'onlineSavingsMsg'
Learn More

Bis(n-propylcyclopentadienyl)magnesium, 98+%, Thermo Scientific™

Catalog Number 15441527
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
114504-74-4
MFCD00151384
magnesium bis 2-propylcyclopenta-1,3-dien-1-ide, magnesium 2+ ion bis 2-propylcyclopenta-1,3-dien-1-ide
This item is not returnable. View return policy
114504-74-4
MFCD00151384
magnesium bis 2-propylcyclopenta-1,3-dien-1-ide, magnesium 2+ ion bis 2-propylcyclopenta-1,3-dien-1-ide

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.94g/mL
Packaging Ampule
Quantity 5 g
UN Number UN3391
Sensitivity Air & Moisture Sensitive
Formula Weight 238.66
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material Bis(n-propylcyclopentadienyl)magnesium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'