missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(mercaptocyclopentane)titanium tetrachloride, 97+%

As a single-source precursor to high quality TiS2 films

Brand:  Alfa Aesar™ 039539.03

Code : NEW

Additional Details : CAS Number : 149391-24-2 Weight : 0.00100kg Transport : UN number : 3261 Chem class : 1a Pack group : II

Quantity 1g

 View more versions of this product

Product Code. 11328338

  • £60.74 / 1g


Estimated Shipment date
from Supplier 19-03-2018
Add to basket

Specifications

Chemical Identifiers

Bis(mercaptocyclopentane)titanium tetrachloride
393.03
UN3261
149391-24-2
MFCD00210605

Specifications

Bis(mercaptocyclopentane)titanium tetrachloride
Crystalline
393.03
UN3261
60°C
Moisture Sensitive
149391-24-2
≥97%
MFCD00210605
Yellow
1g
Videos
SDS