missing translation for 'onlineSavingsMsg'
Learn More

Bis(mercaptocyclohexane)titanium tetrachloride, 99.9% (metals basis), Thermo Scientific™

Catalog Number p-7050814
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
149391-23-1
MFCD00171702
Tetrachlorobis(cyclohexylmercapto)titanium(IV)
This item is not returnable. View return policy
149391-23-1
MFCD00171702
Tetrachlorobis(cyclohexylmercapto)titanium(IV)

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 76°C to 80°C
Color Yellow
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 422.1
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Bis(mercaptocyclohexane)titanium tetrachloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'