missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(isopropylcyclopentadienyl)tungsten dihydride, 98+%

MOCVD precursor

Brand:  Alfa Aesar™ 042672.04

Code : NEW

Additional Details : CAS Number : 64561-25-7 Weight : 0.00200kg Transport : UN number : 3398 Chem class : 1 Pack group : III

Quantity 2g

 View more versions of this product

Product Code. 11325397

  • £181.30 / 2g


Estimated Shipment date
from Supplier 16-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis (isopropylcyclopentadienyl)tungsten dihydride
400.22
UN3398
64561-25-7
MFCD04126431

Specifications

Bis (isopropylcyclopentadienyl)tungsten dihydride
Ampule
≥98%
MFCD04126431
Orange
2g
64561-25-7
Liquid
400.22
UN3398
124°C (0.1mmHg)
Air & Moisture Sensitive
Videos
SDS