missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(ethylcyclopentadienyl)magnesium, 98+%

Brand:  Alfa Aesar™ 038597.03

Code : NEW

Additional Details : CAS Number : 114460-02-5 Weight : 0.00100kg Transport : UN number : 3392 Chem class : 1 Pack group : I

Quantity 1g

 View more versions of this product

Product Code. 15461487

  • £109.40 / 1g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(ethylcyclopentadienyl)magnesium
210.6
UN3392
114460-02-5
MFCD00151375

Specifications

Bis(ethylcyclopentadienyl)magnesium
Ampule
≥98%
MFCD00151375
1.91g/mL
Air & Moisture Sensitive
114460-02-5
Liquid
210.6
UN3392
1g
Videos
SDS