missing translation for 'onlineSavingsMsg'
Learn More

Bis(ethylcyclopentadienyl)magnesium, 98+%, Thermo Scientific™

Catalog Number 15481487
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
114460-02-5
MFCD00151375
This item is not returnable. View return policy
114460-02-5
MFCD00151375

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.91 g/mL
Packaging Ampule
Quantity 250 mg
UN Number UN3392
Sensitivity Air and moisture sensitive
Formula Weight 210.6
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Bis(ethylcyclopentadienyl)magnesium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'