Learn More
Alfa Aesar™ Bis(diphenylphosphino)acetylene, 97%
Brand: Alfa Aesar™ 039277.04
Code : NEW
Additional Details : CAS Number : 5112-95-8 Weight : 0.00200kg
Quantity | 2g |
---|
Chemical Identifiers
Bis(diphenylphosphino)acetylene | |
5112-95-8 | |
394.39 | |
MFCD00003045 |
Specifications
Bis(diphenylphosphino)acetylene | |
Crystalline | |
394.39 | |
85° to 87°C |
5112-95-8 | |
97% | |
MFCD00003045 | |
2g |