missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(dimethylphosphino)methane, 98%

Brand:  Alfa Aesar™ 039275.02

Code : NEW

Additional Details : CAS Number : 64065-08-3 Weight : 0.00050kg Transport : UN number : 2845 Chem class : 1 Pack group : I

Quantity 0.5g

 View more versions of this product

Product Code. 15491527

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Bis(dimethylphosphino)methane
(CH3)2PCH2P(CH3)2
MFCD00058731
64065-08-3
136.11
UN2845

Specifications

Bis(dimethylphosphino)methane
Liquid
(CH3)2PCH2P(CH3)2
MFCD00058731
0.86g/mL
42°C (12mmHg)
Air & Moisture Sensitive
64065-08-3
98%
136.11
UN2845
Colorless
0.5g
Videos
SDS