missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(dimethylamino)chlorophosphine, Thermo Scientific Chemicals
33.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Color | Colorless |
Odor | Pungent |
Linear Formula | [(CH3)2N]2PCl |
Refractive Index | 1.501 |
Quantity | 1 g |
UN Number | UN3265 |
Sensitivity | Air and moisture sensitive |
Formula Weight | 154.58 |
Physical Form | Liquid |
Chemical Name or Material | Bis(dimethylamino)chlorophosphine |
Safety and Handling
Product Identifier
- Bis(dimethylamino)chlorophosphine
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308-IF exposed or concerned:.
- P402-Store in a dry place.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'