missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(diisopropylamino)chlorophosphine, Thermo Scientific Chemicals
189.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
148.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "22914" to get your promotional price.
148.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 100.0°C to 110.0°C |
Packaging | Glass bottle |
Linear Formula | [((CH3)2CH)2N]2PCl |
Quantity | 25 g |
Solubility Information | Solubility in water: reacts |
Formula Weight | 266.8 |
Chemical Name or Material | Bis(diisopropylamino)chlorophosphine |
Safety and Handling
Product Identifier
- Bis(diisopropylamino)chlorophosphine
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'