missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(dibenzylideneacetone)palladium, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 150.0°C |
Color | Black to Brown-Red |
Packaging | Glass bottle |
Linear Formula | Pd(C6H5CH=CHC(O)CH=CHC6H5)2 |
Quantity | 5 g |
Fieser | 15,302 |
Solubility Information | Solubility in water: insoluble. |
Formula Weight | 575.02 |
Percent Purity | 16 to 21% (Pd) |
Physical Form | Fine Powder |
Show More |
Safety and Handling
Product Identifier
- Bis(dibenzylideneacetone)palladium
- Warning
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H290-May be corrosive to metals.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'