Learn More
Alfa Aesar™ Bis(cyclopentadienyl)zirconium dichloride, 99+%
Brand: Alfa Aesar™ 039558.03
Code : NEW
Additional Details : CAS Number : 1291-32-3 Weight : 0.00100kg
Quantity | 1g |
---|
Description
- Packaged under argon
Chemical Identifiers
Bis(cyclopentadienyl)zirconium dichloride | |
292.32 | |
Zirconocene dichloride; Dichlorobis(cyclopentadienyl)zirconium |
1291-32-3 | |
MFCD00003726 |
Specifications
Bis(cyclopentadienyl)zirconium dichloride | |
Powder | |
292.32 | |
White to off-white | |
124° to 125°C (15mmHg) | |
Zirconocene dichloride; Dichlorobis(cyclopentadienyl)zirconium |
1291-32-3 | |
≥99% | |
MFCD00003726 | |
242° to 245°C | |
1g | |
Air & Moisture Sensitive |