missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(cyclopentadienyl)zirconium dichloride, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
Zirconocene dichloride can also be used to prepare the Negishi reagent, Cp2Zr(η2-butene) which is used as a source for Cp2Zr in oxidative cyclisation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
Melting Point | 242°C to 245°C |
Color | White |
Boiling Point | 124°C to 125°C (15 mmHg) |
Quantity | 1 g |
Sensitivity | Air and moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 292.32 |
Percent Purity | ≥99% |
Physical Form | Powder |
Chemical Name or Material | Bis(cyclopentadienyl)zirconium dichloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'