missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(cyclopentadienyl)vanadium dichloride, 95%

Brand:  Alfa Aesar™ 039588.06

Code : NEW

Additional Details : CAS Number : 12083-48-6 Weight : 0.00500kg Transport : UN number : 3285 Chem class : 1 Pack group : II

Quantity 5g

 View more versions of this product

Product Code. 11304507

  • £315.10 / 5g


Estimated Shipment date
from Supplier 14-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(cyclopentadienyl)vanadium dichloride
252.04
UN3285
12083-48-6
MFCD00014260
Vanadocene dichloride

Specifications

Bis(cyclopentadienyl)vanadium dichloride
Pungent
95%
252.04
UN3285
250°C (dec.)
Vanadocene dichloride
12083-48-6
Crystalline
1.60g/mL
MFCD00014260
Green
5g
Air & Moisture Sensitive
Videos
SDS