missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)molybdenum dichloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7050750
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy
12184-22-4
MFCD00009948
This item is not returnable. View return policy
12184-22-4
MFCD00009948

Bis(cyclopentadienyl)molybdenum dichloride, 99% is used to prepare many derivatives. It is also used as pharmaceutical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 0.5 g
UN Number UN3261
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 297.04
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)molybdenum dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'