missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(Boc-aminooxy)acetic acid, 98%

Brand:  Alfa Aesar™ H62879.03

Code : NEW

Additional Details : CAS Number : 293302-31-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15469328

  • £75.30 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Description

Description

Applications
Bis(Boc-aminooxy)acetic acid employed as a protecting reagent and intermediate pharmaceuticals and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

Bis(Boc-aminooxy)acetic acid
[(CH3)3COCONHO]2CHCOOH
MFCD06410966
293302-31-5
322.32
Bis-Boc-Aoa

Specifications

Bis(Boc-aminooxy)acetic acid
Powder
[(CH3)3COCONHO]2CHCOOH
MFCD06410966
1g
293302-31-5
98%
322.32
White
Bis-Boc-Aoa
Videos
SDS