missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetonitrile)palladium(II) chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-68033
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
14592-56-4
C4H6Cl2N2Pd
259.42
MFCD00013122
XGPJSRXOBCGBMN-UHFFFAOYSA-L
palladium(2+) bis(acetonitrile) dichloride
CC#N.CC#N.Cl[Pd++]Cl
14592-56-4
C4H6Cl2N2Pd
259.42
MFCD00013122
XGPJSRXOBCGBMN-UHFFFAOYSA-L
palladium(2+) bis(acetonitrile) dichloride
CC#N.CC#N.Cl[Pd++]Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 300°C
Color Brown-Rust to Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3CN)2PdCl2
Quantity 500 mg
Fieser 13,33; 15,28; 16,25; 17,30; 01,1015; 03,254; 04,435
Solubility Information Solubility in water: insoluble
Formula Weight 259.4
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Bis(acetonitrile)palladium(II) chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(acetonitrile)palladium(II) chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'