missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(8-hydroxyquinolinato)zinc, 98%

As a photoluminescent (PL) material; epsilon(PL) = 556nm

Brand:  Alfa Aesar™ 043850.MD

Code : NEW

Additional Details : CAS Number : 13978-85-3 Weight : 0.01000kg

Quantity 250mg

 View more versions of this product

Product Code. 11340518

  • £41.23 / 250mg


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

Bis (8-hydroxyquinolinato)zinc
C18H12N2O2Zn
MFCD00065291
13978-85-3
353.69
8-Hydroxyquinolinezinc; Zinc 8-hydroxyquinolinate

Specifications

Bis (8-hydroxyquinolinato)zinc
13978-85-3
Yellow
98%
353.69
8-Hydroxyquinolinezinc; Zinc 8-hydroxyquinolinate
151°C
250mg
Powder
C18H12N2O2Zn
MFCD00065291
Videos
SDS