missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate, Electronic grade, ≥99% (metals basis)

Brand:  Alfa Aesar™ H55976.03

Code : NEW

Additional Details : CAS Number : 84563-54-2 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15455295

  • £87.66 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate
Electrophoresis Grade
MFCD02683471
84563-54-2
542.4
Bis(4-tert-butylphenyl)iodonium triflate

Specifications

Bis(4-tert-butylphenyl)iodonium trifluoromethanesulfonate
Electrophoresis Grade
≥99%
542.4
Bis(4-tert-butylphenyl)iodonium triflate
162° to 166°C
Light Sensitive
84563-54-2
Powder
(metals basis)
MFCD02683471
White
1g
Videos
SDS