missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate, Electronic grade, ≥99% (metals basis), Thermo Scientific Chemicals

Catalog Number 15475285
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
131717-99-2
C27H33IO3S
564.52
MFCD02683472
UEJFJTOGXLEPIV-UHFFFAOYSA-M
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis
16216932
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1
This item is not returnable. View return policy
131717-99-2
C27H33IO3S
564.52
MFCD02683472
UEJFJTOGXLEPIV-UHFFFAOYSA-M
bis 4-tert-butylphenyl iodanium tosylate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, 4,4-di-tert-butyldiphenyliodonium p-toluenesulfonate, acmc-20n1fr, bis 4-tert-butylphenyl iodonium tosylate, bis 4-tert-butylphenl iodonium p, bis p-tert-butylphenyl iodonium p-toluenesulfonate, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade, bis 4-tert-butylphenyl iodonium p-toluenesulfonate, electronic grade trace metals basis
16216932
bis(4-tert-butylphenyl)iodanium;4-methylbenzenesulfonate
CC1=CC=C(C=C1)S([O-])(=O)=O.CC(C)(C)C1=CC=C([I+]C2=CC=C(C=C2)C(C)(C)C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 237°C to 240°C
Color White
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 564.52
Percent Purity ≥99%
Grade Electronic
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(4-tert-butylphenyl)iodonium p-toluenesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'