missing translation for 'onlineSavingsMsg'
Learn More

Bis-(4-hydroxy-1-naphthyl)phenylmethanol, ACROS Organics™

Brand:  Acros Organics 301640050

Code : 29

Additional Details : CAS Number : 6948-88-5 Weight : 0.05500kg

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10541521

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

Bis-(4-hydroxy-1-naphthyl)phenylmethanol
C27H20O3
MFCD00003979
6948-88-5
392.46
α-Hydroxy-α-(4-hydroxy-1-naphthalenyl)-α-phenyl-1-naphthalenemethanol, 4, 4'-(α-Hydroxybenzylidene)di-1-naphthol

Specifications

Bis-(4-hydroxy-1-naphthyl)phenylmethanol
Authentic
Powder
350 min. (648nm, 0.01N NaOH)
MFCD00003979
from pH 8.2 (orange) to pH 10.0 (bluish-green)
Red-brown
5g
6948-88-5
Glass bottle
C27H20O3
392.46
06, IV, 7666
α-Hydroxy-α-(4-hydroxy-1-naphthalenyl)-α-phenyl-1-naphthalenemethanol, 4, 4'-(α-Hydroxybenzylidene)di-1-naphthol
121.0° to 125.0°C
Videos
SDS