missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%, ACROS Organics™

Brand:  Acros Organics 310060050

Code : 29

Additional Details : CAS Number : 68641-49-6 Weight : 0.05500kg Transport : UN number : 1759 Chem class : 1a Pack group : II

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10532851

  • £27.00 / 5g


EU Stock 5
Add to basket

Specifications

Chemical Identifiers

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
96.0
≥96.0%
C6H8ClN2O5P
MFCD00010077
68641-49-6
100.0
97%
254.57

Specifications

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride
96.0
Authentic
Crystalline Powder
97%
254.57
White to off-white
68641-49-6
100.0
Glass bottle
≥96.0%
C6H8ClN2O5P
MFCD00010077
5g
Videos
SDS