missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-methoxyethyl) phthalate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
Description
It finds it application in cosmetic industry. It is also used as a plasticizer for toys, including inflatable water products, hoppers, play and exercise balls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 1.17 g/mL |
Boiling Point | 340°C |
Flash Point | 185°C (365°F) |
Refractive Index | 1.503 |
Quantity | 50 g |
Beilstein | 2056929 |
Solubility Information | Soluble in water 8500mg/L at 25°C. |
Formula Weight | 282.29 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'