missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-methoxyethyl) ether, 99%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -64.0°C |
Density | 0.9370g/mL |
Boiling Point | 162.0°C |
Flash Point | 51°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3OCH2CH2)2O |
Refractive Index | 1.4070 to 1.4090 |
Quantity | 2.5 L |
Show More |
Safety and Handling
Product Identifier
- 2-Methoxyethyl ether
- Danger
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- H226-Flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'