missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-diphenylphosphinophenyl)ether, 99%, ACROS Organics™

Brand:  Acros Organics 383370050

Code : 29

Additional Details : CAS Number : 166330-10-5 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10524103

  • £39.20 / 5g


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

Bis(2-diphenylphosphinophenyl)ether
98.5
99%
C36H28OP2
538.56
166330-10-5
100.0
99%
[(C6H5)2P(C6H4)]2O
MFCD00233863

Specifications

Bis(2-diphenylphosphinophenyl)ether
98.5
Authentic
Crystalline Powder or Crystals
99%
[(C6H5)2P(C6H4)]2O
MFCD00233863
184.0° to 187.0°C
166330-10-5
100.0
Glass bottle
99%
C36H28OP2
538.56
White to off-white
5g
Videos
SDS