missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(diisopropylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals

Catalog Number p-7040647
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
131890-26-1
C16H37NP2
305.427
MFCD17018778
FTVIGQGOGIHMBS-UHFFFAOYSA-N
bis 2-di-i-propylphosphino ethyl amine, di 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphanyl ethyl amine, bis 2-diisopropylphosphino ethyl amine solution, 10 wt. % in thf
15757160
2-di(propan-2-yl)phosphanyl-N-[2-di(propan-2-yl)phosphanylethyl]ethanamine
CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C
This item is not returnable. View return policy
131890-26-1
C16H37NP2
305.427
MFCD17018778
FTVIGQGOGIHMBS-UHFFFAOYSA-N
bis 2-di-i-propylphosphino ethyl amine, di 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphino ethyl amine, bis 2-diisopropylphosphanyl ethyl amine, bis 2-diisopropylphosphino ethyl amine solution, 10 wt. % in thf
15757160
2-di(propan-2-yl)phosphanyl-N-[2-di(propan-2-yl)phosphanylethyl]ethanamine
CC(C)P(CCNCCP(C(C)C)C(C)C)C(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis[2-(diisopropylphosphino)ethyl]amine acts as ligand and form complexes with ruthenium, which finds application in the dehydrogenation of ethanol.

Solubility
Immiscible with water.

Notes
Air sensitive. Incompatible with oxidizing agents and oxygen.
TRUSTED_SUSTAINABILITY
Density 0.884
Linear Formula ([(CH3)2CH]2PCH2CH2)2NH
Refractive Index 1.418
Quantity 1 g
UN Number UN2924
Sensitivity Air Sensitive
Solubility Information Immiscible with water.
Formula Weight 305.42
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(diisopropylphosphino)ethyl]amine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'