missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane, +95%, ACROS Organics™

Brand:  Acros Organics 388901000

Code : 29

Additional Details : CAS Number : 89597-05-7 Weight : 0.00010kg

Quantity 100mg
Packaging Glass bottle

 View more versions of this product

Product Code. 10320804

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
95.0
≥95%
C20H32Si
MFCD00674017
89597-05-7
100.0
95+%
300.56

Specifications

Bis(2,3,4,5-tetramethyl-2,4-cyclopentadiene-1-yl)dimethylsilane
95.0
Authentic
Powder and Chunks
95+%
300.56
Yellow
100mg
89597-05-7
100.0
Glass bottle
≥95%
C20H32Si
MFCD00674017
65.0° to 68.0°C
Videos
SDS