missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc(II), 99.9% (metals basis)

Chemical vapor deposition of Zinc oxide

Brand:  Alfa Aesar™ 043037.03

Code : NEW

Additional Details : CAS Number : 14363-14-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11324688

  • £36.68 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Bis (2,2,6,6-tetramethyl-3,5-heptanedionato)zinc (II)
C22H38O4Zn
MFCD00192578
14363-14-5
431.93
Zinc-DPM

Specifications

Bis (2,2,6,6-tetramethyl-3,5-heptanedionato)zinc (II)
14363-14-5
250°C (dec.)
99.9%
C22H38O4Zn
MFCD00192578
144°C
1g
Crystalline
(metals basis)
431.93
Zinc-DPM
Videos
SDS