Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)zinc(II), 99.9% (metals basis)

Chemical vapor deposition of Zinc oxide

Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 043037.06

BIS(2266-TETRAMETHYL-35- HEPT.)ZINC(II) 5G

UNSPSC: 12352103

Code: NEW

Additional Details:
Additional Details: CAS Number: 14363-14-5 Weight: 0.01000kg


Quantity 5g

Product Code. 11334688

Quantity Price
1 £ 140.87 / 5g
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material Bis (2,2,6,6-tetramethyl-3,5-heptanedionato)zinc (II)
CAS 14363-14-5
Molecular Formula C22H38O4Zn
Formula Weight 431.93
MDL Number MFCD00192578
Synonym Zinc-DPM

Specification

Formula Weight 431.93
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Bis (2,2,6,6-tetramethyl-3,5-heptanedionato)zinc (II)
Melting Point 144°C
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (metals basis)
Molecular Formula C22H38O4Zn
MDL Number MFCD00192578
CAS 14363-14-5
Synonym Zinc-DPM
Health Hazard 1 Warning
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 250°C (dec.)
Safety and Handling

Warning

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.