missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium(II), 99.99% (metals basis)

MOCVD preparation of thin films containing strontium

Brand:  Alfa Aesar™ 041608.03

Code : NEW

Additional Details : CAS Number : 36830-74-7 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 11368748

  • £37.74 / 1g


Estimated Shipment date
from Supplier 28-02-2018
Add to basket

Specifications

Chemical Identifiers

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium(II)
454.16
Strontium-DPM; Strontium(II) bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
36830-74-7
MFCD00064758

Specifications

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium(II)
Powder
(metals basis)
MFCD00064758
200°C
1g
36830-74-7
99.99%
454.16
Strontium-DPM; Strontium(II) bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
230°C (0.7mmHg) (subl.)
Videos
SDS