missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II), Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 126°C to 128°C |
Boiling Point | 134°C (0.1 mmHg)(sublimation) |
Quantity | 1 g |
UN Number | UN3467 |
Formula Weight | 573.75 |
Physical Form | Powder |
Chemical Name or Material | Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II) |
Safety and Handling
Product Identifier
- Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)lead(II)
- Danger
- Acute toxicity Category 4
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'