missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.
Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications
Density | 1.545 g/mL |
Boiling Point | 43°C to 44°C (2 mmHg) |
Flash Point | 76°C (169°F) |
Linear Formula | (CF3CH2O)2P(O)H |
Refractive Index | 1.332 |
Quantity | 5 g |
Formula Weight | 246.05 |
Percent Purity | 90% |
Grade | Technical |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'