missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl)amine, 95%, Thermo Scientific Chemicals

Catalog Number p-7046892
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
407-01-2
C4H5F6N
181.081
MFCD00042094
GTJGHXLFPMOKCE-UHFFFAOYSA-N
78999
2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine
C(C(F)(F)F)NCC(F)(F)F
This item is not returnable. View return policy
407-01-2
C4H5F6N
181.081
MFCD00042094
GTJGHXLFPMOKCE-UHFFFAOYSA-N
78999
2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine
C(C(F)(F)F)NCC(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.1
Boiling Point 70°C to 71°C
Refractive Index 1.301
Quantity 1 g
UN Number UN2735
Beilstein 1811926
Synonym bis 2,2,2-trifluoroethyl amine, bis trifluoroethyl amine, 1,1,1,5,5,5-hexafluoro-3-azapentane, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl ethanamine, ethanamine, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl, bis trifluoroethyl-amin, acmc-1aha1, 1,1,15,5,5-hexafluoro-3-azapentane, ethanamine,2,2,2-trifluoro-n-2,2,2-trifluoroethyl, 2,2,2-tris fluoranyl-n-2,2,2-tris fluoranyl ethyl ethanamine
Formula Weight 181.08
Percent Purity 96%
Chemical Name or Material Bis(2,2,2-trifluoroethyl)amine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'