missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(1H-inden-1-yl)dimethylsilane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Yellow to Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Molecular Formula | C20H20Si |
Quantity | 1 g |
Molecular Weight (g/mol) | 288.465 |
Formula Weight | 288.46 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'