missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, Thermo Scientific Chemicals

Catalog Number p-3212421
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
35138-22-8
C16H24BF4Rh
406.08
MFCD00075045
NBGSCPNNKGVTKU-QMDOQEJBSA-N
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
5702662
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
35138-22-8
C16H24BF4Rh
406.08
MFCD00075045
NBGSCPNNKGVTKU-QMDOQEJBSA-N
bis 1,5-cyclooctadiene rhodium i tetrafluoroborate, rhodium i tetrafluoroborate 1,5-cyclooctadiene complex, 1z,5z-cycloocta-1,5-diene; rhodium; tetrafluoroborate, bis 1,5-cyclooctadiene rhodium tetrafluoroborate, rh cod 2 bf4, c16h24bf4rh, c16h24rh.bf4, bis 1,5-cyclooctadien rhodium i tetrafluoroborate
5702662
(1Z,5Z)-cycloocta-1,5-diene;rhodium;tetrafluoroborate
[Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 210.0°C
Color Red
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula [Rh(cod)2]BF4
Quantity 250 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons
Formula Weight 406.08
Physical Form Powder
Chemical Name or Material Bis(1, 5-Cyclooctadiene)rhodium(I) tetrafluoroborate
compliance-icons compliance-icons
Product Identifier
  • Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'