missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate, Thermo Scientific Chemicals

Catalog Number p-7059872
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
130296-28-5
C16H24F6RhSb
555.02
MFCD11113031
BROZUPPJWNODCU-SXGOCSEPSA-H
bis 1,5-cyclooctadiene rhodium i hexafluoroantimonate
16689296
[Rh+].F[Sb-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
This item is not returnable. View return policy
130296-28-5
C16H24F6RhSb
555.02
MFCD11113031
BROZUPPJWNODCU-SXGOCSEPSA-H
bis 1,5-cyclooctadiene rhodium i hexafluoroantimonate
16689296
[Rh+].F[Sb-](F)(F)(F)(F)F.C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Brown
Quantity 250 mg
UN Number UN3077
IUPAC Name cycloocta-1,5-diene;hexafluoroantimony(1-);rhodium
Formula Weight 555.01
Physical Form Crystalline Powder
Chemical Name or Material Bis (1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'