missing translation for 'onlineSavingsMsg'
Learn More

Biperiden hydrochloride, Tocris Bioscience™

Muscarinic receptor antagonist; some selectivity for M1

Brand:  Tocris (Bio-Techne) 285350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13441944

  • £336.61 / 50mg

Estimated Shipment date
from Supplier 01-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Muscarinic receptor antagonist that displays some selectivity for the M1 subtype (Ki values are 0.48, 2.4, 3.9, 6.3 and 6.3 nM for M1, M4, M3, M2 and M5 receptors respectively). Alleviates extrapyramidal symptoms associated with antipsychotic drugs. Displays antiparkinsonian activity.
Specifications

Chemical Identifiers

1235-82-1
347.927
1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
(1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
C21H30ClNO
RDNLAULGBSQZMP-LIJKWCKLSA-N
129318274
C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl

Specifications

Biperiden hydrochloride
1235-82-1
1r-1-1r,4r,5s-5-bicyclo 2.2.1 hept-2-enyl-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
C1CCN(CC1)CCC(C2CC3CC2C=C3)(C4=CC=CC=C4)O.Cl
347.927
347.92
50mg
C21H30ClNO
RDNLAULGBSQZMP-LIJKWCKLSA-N
(1R)-1-[(1R,4R,5S)-5-bicyclo[2.2.1]hept-2-enyl]-1-phenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride
129318274
>99%
Videos
Documents