Learn More
Biotin-Anti-His-Tag Antibody
Biotin-Anti-His-Tag Antibody
Brand: Invitrogen PV6090
Code : 50
Additional Details : Weight : 0.50000kg
Description
Biotin-labeled anti-His-tag antibodies are used to indirectly label proteins tagged with 6XHis. This can be advantageous when working with partially purified protein, or in cases in which direct labeling of the protein using biotin negatively impacts protein activity. These antibodies can be used with our Kinase Tracers and Eu-Streptavidin to conduct LanthaScreen™ Eu Kinase Binding Assays. Learn More and See Which Kinases Have Been Validated Using This AssaySpecifications
Kinase Assay, Kinase Assay | |
Biotin | |
150kDa | |
RUO, RUO | |
Primary | |
Biotin-labeled anti-His tag antibody in HEPES buffered saline. Store at -20°C. | |
Dry Ice |
0.5 mg⁄ml | |
Biotin-Anti-His-Tag Antibody, 1mg, 0.5mg/mL Concentration, Biotin Conjugate, HEPES buffered saline (137mM NaCl, 2.7mM KCl, 10mM HEPES pH 7.5) Storage Buffer, Used for Pharma and Biopharma, Target and Lead Identification and Validation | |
1 mg | |
Kinase Biology | |
1 Each | |
Frozen |
For Research Use Only. Not for use in diagnostic procedures.