Learn More
Agilent BioTek Waste Bottle and Tubing
For use with 405 LS , 405 Touch, EL406, ELx405
Brand: Agilent BioTek 7100556
Additional Details : Weight : 0.10000kg
Description
Cap sold separately. (See Cat. No. 12-566-720)
Specifications
20 L | |
676.28 oz. | |
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 4 L or 10 L Bottles Complete Waste System |
Waste Bottle | |
Tubing |