Learn More
Synergy™ NEO HTS Multi-Mode Microplate Readers
With multiple parallel detectors for ultra-fast measurements and a dedicated filter-based system for live cell assays
Brand:  Biotek Instruments NEOALPHAB
Code : Z1
Additional Details : Weight : 35.00000kg
 View more versions of this product
Product Code. 12847142
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Multi-Mode Microplate Reader
Xenon flash
35kg
With Alpha laser, fluorescence intensity, fluorescence polarization, time resolved fluorescence, UV-Vis absorbance, luminescence
Videos