Learn More
BioTek™ Synergy™ NEO HTS Multi-Mode Microplate Readers
With multiple parallel detectors for ultra-fast measurements and a dedicated filter-based system for live cell assays
Brand:  BioTek™ NEOB
Code : Z1
Additional Details : Weight : 35.00000kg
 View more versions of this product
Product Code. 12817142
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Specifications
Specifications
Multi-Mode Microplate Reader
96 wells: 6 seconds; 384 well: 11 seconds; 1536 well: 25 seconds
With fluorescence intensity, fluorescence polarization, time resolved fluorescence, UV-Vis absorbance, luminescence
Xenon flash
35kg
Videos