missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Synergy™ HTX Multi-Mode Microplate Reader

Compact, affordable system automates absorbance, fluorescence, luminescence and AlphaScreen™/AlphaLISA™ measurements. BioTek Synergy HTX Multi-Mode Microplate Reader provides superior performance in all detection modes using a unique dualoptics design. The absorbance detection optics design uses a xenon flash lamp and monochromator for filter-free wavelength selection from 200 to 999nm in 1nm increments.

Brand:  BioTek™ S1L

Additional Details : Weight : 18.00000kg

 View more versions of this product

Product Code. 15346166

  • £11880.00 / Each


Estimated Shipment date
from Supplier 25-04-2019
Add to basket

Description

Description

Fluorescence determinations are made using a tungsten halogen lamp with interference filters in conjunction with a PMT detector for maximum sensitivity. For increased workflow automation and throughput, BioTek's BioStack can be easily connected to Synergy HTX to automatically process up to 50 microplates at a time.

  • For 6- to 384-well microplates and Take3 microvolume plates
  • Flexibility of monochromator based absorbance with high performance filter-based fluorescence/luminescence
  • 2μL low volume nucleic acid quantification with Take3 and Take3 Trio plates
  • Cell friendly orbital shaking and advanced incubator design to 50°C with top/bottom gradient minimizing plate lid condensation
  • Dual reagent injectors for inject/read applications, such as enzyme kinetics and Dual-Luciferase™ Reporter
  • AlphaScreen™/AlphaLISA™
  • Modular and upgradable
  • Unique 4-Zone™ incubation to 50°C
  • Advanced shaking profiles including linear and orbital
  • Compatible with BioStack™ and third party automation Scan
  • Powerful Gen5™ Software for data collection, analysis, exporting and reporting
Specifications
  • Detection method: Fluorescence, Time-Resolved Fluorescence (secondary mode), Luminescence, UV-Visible Absorbance
  • Read method: End point, kinetic, spectral scanning, well-area scaning
  • Microplate types: 6 to 384 wells, PCR plates, Take3 and Take3 Trio Micro-Volume Plates
  • Temperature control: 4°C above ambient to 50°C; ±0.2°C at 37°C

Applications
Nucleic acid quantification, protein quantification, enzyme kinetics, biomarker quantification, ELISAs, genetic analysis, cell proliferation, cytotoxicity, drug absorption and metabolism, food safety, and environmental monitoring.

Order Info

Optional Accessories: Dual Reagent Dispenser Module, Gen5 Secure (for 21 CFR Part 11 Compliance), Fluorescence Test Plate, Absorbance Test Plate, Luminescence Test Plate, Patented BioCell™ 1 cm quartz vessel, Product Qualification Package, Take3/Take3 Trio.

Compliance

CE and TUV marked

Specifications

Specifications

Multi-Mode Microplate Reader
0-4.0 OD/>6 decades
18kg
Luminescence
Fluorescence, Time-Resolved Fluorescence (secondary mode), Luminescence, UV-Visible Absorbance
1 USB, 1 RS232 for external PC control
100-240V AC 50/60 Hz
Videos