missing translation for 'onlineSavingsMsg'
Learn More

BioTek™ Synergy™ H1m Monochromator-Based Multi-Mode Microplate Readers

Upgradeable to patented Hybrid monochromator-filter optical system.

Brand:  BioTek™ H1MDG

Additional Details : Weight : 22.50000kg

 View more versions of this product

Product Code. 15356156

  • £29300.00 / Each

Estimated Shipment date
from Supplier 02-05-2018
Add to basket



  • Fully monochromator-based with quadruple grating architecture
  • Compatible with Take3 Micro-Volume Plate: sixteen 2μL microspots for low volume DNA/RNA 260 nm measurements
  • High performance fluorescence (top and bottom), absorbance (UV visible), TRF and luminescence detection systems accommodate all life-science research microplate applications
  • Automated Z-height adjustment accommodates variable microplate heights and liquid levels with 100μm resolution
  • Optional dual-reagent dispenser can be used for assays requiring precise timing
  • Comes with Gen5™ software: reader control, advanced data analysis and flexible Excel export in one software package
  • Accomodates 1 to 384 well microplates
  • Automation: Yes

H1MG and H1MDG

  • Gas controller compatible
  • Gas module must be purchased separately (BT1210007 or BT12310008)


CE marked



Multi-Mode Microplate Reader
With dual reagent dispenser
Gen5 Data Analysis Software